Would you like to log in again?
Clip-On Welders Adapter, MSA
Supplier: MSA
454-10077063
300080-966EA 405.21 USD InStock
300080-966
Clip-On Welders Adapter, MSA
Welding Supplies
Clip-On Welder's Adapter for Advantage 4200 Twin Port Face Piece
Supplied with cover lens and lens filter plate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty