Would you like to log in again?
Powerlock® Tape Rules, ³/₄" Wide Blade, Stanley® Products
Supplier: Stanley
Powerlock®
680-33-116
300009-898EA 75.5 USD InStock
300009-898
Powerlock® Tape Rules, ³/₄" Wide Blade, Stanley® Products
Measuring Tapes
  • 7' blade standout
  • Mylar® polyester film extends life of entire blade
  • Chrome high-impact ABS case is job site tough
  • Secure blade lock won't creep during measurements
  • Corrosion-resistant Tru-zero end hook for accurate measurements
Single scale, with measurements in inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty