Would you like to log in again?
Infant Heel Warmer, Azer Scientific
Supplier: Azer Scientific
ES23532 ES23532
10836-040CS 160.7 USD InStock
10836-040 CA11012-546
Infant Heel Warmer, Azer Scientific
First Aid Supplies Surgical Drapes
Safely and effectively warms infant heel for capillary blood collection.
  • Heats to an optimal temperature of 104 °F
  • Liquid filled pouch conforms to infant heel for even heat distribution
  • Attached strap secures device to foot
The Infant heel warmer safely and comfortably increases blood flow to the infant heel area to assist with capillary blood collection. The 5×3.5" liquid filled pouch features a trigger disc inside for quick activation, is non-toxic and latex free.
Packaging: 25/pack, 4 packs/case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty