Would you like to log in again?
Mounting Medium with Toulene, Azer Scientific
Supplier: Azer Scientific
ES725 ES726
10821-476CS 397.26 USD InStock
10821-476 10821-478
Mounting Medium with Toulene, Azer Scientific
Stains Microscopy and staining reagents
Mounting Medium with Toluene is used for both mounting and long-term storage of slides.
  • Ideal for mounting and long-term storage of slides
  • Provides thinner adhesive layer for better optical quality
  • Low viscosity
  • 4 oz. dropper bottle offers easy application, whereas 16 oz. bottle is ideal for labs with high throughput
This traditional mounting medium is compatible with xylene. It provides protection from photobleaching and fading of stains.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty