Would you like to log in again?
ODN M383 (TLRgrade™) (synthetic) (Control), Enzo Life Sciences
Supplier: Enzo Life Sciences
746-204-T100
89153-108EA 158.5 USD InStock
89153-108
ODN M383 (TLRgrade™) (synthetic) (Control), Enzo Life Sciences
Nucleic Acid Reagents Primers and Probes
Negative control for ODN M362
Synthetic.
GpC oligodeoxynucleotide with phosphorothioate backbone.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty