Would you like to log in again?
Chemkey® Reagents for SL1000 Portable Parallel Analyzer (PPA)
Supplier: Hach
9429400 9429600 9429200 9436800 8636000 8636400 8636100 8499400 8499300 9429000 8636600 8636500 9425200 8791900 9429300 9427900 9429100 8636200 8636300 9429500
76196-632BX 47.94 USD InStock
76196-632 76196-620 76196-652 76196-674 76196-630 76196-662 76196-672 76196-650 76196-670 76196-628 76196-638 76196-626 76196-648 76196-658 76196-646 76196-624 76196-656 76196-634 76196-666 76196-644
Chemkey® Reagents for SL1000 Portable Parallel Analyzer (PPA)
Environmental Test Kits
Use Hach's patented Chemkey™ technology with the SL1000 Portable Parallel Analyzer (PPA) for dramatically streamlined water quality testing
Chemkeys are a new technology that are designed for use with the SL1000-PPA Portable Parallel Analyzer.
Same Trusted Technology: Chemkey® reagents execute the same process steps that you have trusted for decades — now delivered in a simple, self-contained package. All chemicals and processes are entirety contained inside the Chemkey®.
Less Hassle: There is no zeroing, no mixing, no shaking, no chemicals or vials to handle. All you have to do is insert the chemkey and let the PPA-SL1000 read you back the results.
Increasing Portfolio: So far, we have released Chemkeys in 3 different phases. We initially released our Phase I Chemkeys which are: monochloramine, free and total ammonia, free and total chlorine, nitrite, copper. The following Phase II Chemkeys have been released: orthophosphate, hardness, alkalinity, and dissolved iron. Phase III parameters include: Extended range chlorine (up to 10 mg/L), Combined Free and Total Ammonia (two measurements, one chemkey), and High Range Hardness.
Certifications: Chlorine Chemkeys are USEPA accepted for reporting in drinking water analysis using Hach Method 10260.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty