Would you like to log in again?
Riboprobe System Buffers, Promega
Supplier: Promega
P1121
PAP1121EA 139.5 USD InStock
PAP1121
Riboprobe System Buffers, Promega
Nucleic Acid Reagents RNA Synthesis and Transcription
Riboprobe System Buffers are qualified for use in applications where maintaining the integrity of RNA is critical. Product is quality tested to ensure the absence of detectable RNase activity.
  • Package of Transcription-Optimized Buffer, rNTPs and Nuclease-Free Water
  • Used with Riboprobe(R) Systems
  • Qualified for use for in vitro transcription reactions with SP6, T7 or T3 RNA polymerase
  • Reagents tested with other Riboprobe(R) System components. rNTPs tested for functionality with in vitro transcription reactions.
Riboprobe System Buffers are components of the single and combination Riboprobe Systems. The buffers also are available as standalone products. RQ1 RNase-Free DNase is used to remove template DNA from RNA preparations and is qualified for use in applications where maintaining the integrity of RNA is critical. Product is quality tested to ensure the absence of detectable RNase activity. 10X Reaction Buffer and 10X Stop Buffer included. rATP, rCTP, rGTP and rUTP are provided in individual tubes, qualified for use with the Riboprobe Systems. The rNTPs are supplied in nuclease-free water. Purity has been verified by HPLC analysis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty