Would you like to log in again?
RVprimer4 (counterclockwise), 2 µg, Promega
Supplier: Promega
E4491
PAE4491EA 97.65 USD InStock
PAE4491
RVprimer4 (counterclockwise), 2 µg, Promega
Nucleic Acid Reagents Sanger Sequencing Reagents
The Reporter Vector (RV) Sequencing Primers are designed for sequencing inserts in the pGL3, pGL4, Chroma-Luc and pCAT3 Reporter Vectors.
  • Designed for Use with the pGL4 Luciferase Vectors and Other Luciferase and CAT Reporter Vectors
  • Sequence double-stranded templates using both primers
  • Sequence single-stranded templates only using only RVprimer4
The Reporter Vector (RV) Sequencing Primers are designed for use with the pGL3 and pGL4 Luciferase Vectors, Chroma-Luc Vectors and pCAT3 Reporter Vectors. RVprimer3 binds upstream of the luc+, luc2 or CAT gene, and sequencing runs clockwise across the multiple cloning region. RVprimer4 binds downstream of the luc+, luc2 or CAT polyadenylation region in the Promoter and Basic Vectors and downstream of the SV40 enhancer region of the Enhancer and Control Vectors. Both primers can be used for sequencing double-stranded templates, but only RVprimer4 can be used for sequencing single-stranded templates. Primer Sequences: RVprimer3, 5'-d(CTAGCAAAATAGGCTGTCCC)-3'; RVprimer4, 5'-d(GACGATAGTCATGCCCCGCG)-3'.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty