Would you like to log in again?
T3 and T7 Polymerases, Agilent Technologies
Supplier: Agilent Technologies
600111 600123 600124 600111 600124 600123
ST600123EA 137.54 USD InStock
ST600123 99902-278 ST600111 CA99902-278 CAST600111 CAST600123
T3 and T7 Polymerases, Agilent Technologies
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
DNA-dependent RNA polymerases with high sequence specificity for T3 or T7 promoters.
  • Probes for nucleic acid hybridizations
  • Templates for in vitro translations
  • Substrates for RNA processing studies
  • Exon and intron mapping of genomic DNA
Bacteriophage T3 and T7 RNA polymerases are DNA-dependent RNA polymerases with high sequence specificity for T3 or T7 promoters. Both T3 and T7 RNA polymerases synthesize RNA 5' to 3' and incorporate 35S, 32P, and 33P ribonucleotides. These enzymes are isolated from an overproducing recombinant E. coli clone.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty