Would you like to log in again?
PathDetect Cis-Reporting Systems, Agilent Technologies
Supplier: Agilent Technologies
240120 219074 240116 219074 219085 240131 240053 240051 219092 240130 219076 219059 219075 219071 240133 219091 219083 240039 219094 219087 219088 240119 219082 219077 240129 219084 219075 240115 219091 240113 219090 219071 240114 240112 240111 219095 219076 219078 219089 219073 219093 240050 219089 240049 219079 219080 219094 240051 240049 219077 240134 219087 240050 219078 240136 240053 240132 240135 240134 219081
99900-686EA 408.74 USD InStock
CA99900-974 CA99900-696 99900-686 CA99900-698 CA99900-730 99900-688 CA99900-818 CA99900-978 CA99900-736 CA99900-816 CA99900-738 CA99900-780 99900-692 CA99900-820 CA99900-988 CA99900-822 99900-736 CA99900-982 CA99900-984 97066-352 97066-354 95040-796 99900-816 CA99900-824 CA99900-706 97066-350 CA99900-990 99900-780 97066-348 99900-822 CA99900-996 CA99900-636 CA99900-998 CA95040-758 CA99900-674 CA95040-796 CA97066-354 CA99900-994 CA99900-718 CA97066-350 CA97066-352 CA95040-750 CA99901-000 CA99900-680 CA99900-682 CA95040-748 CA95040-744 CA99900-688 CA95040-746 CA97066-348 CA99900-720 CA99900-686 CA99900-806 CA99900-728 99901-000 CA99901-004 CA99901-002 CA99900-692 99900-682 99900-680
PathDetect Cis-Reporting Systems, Agilent Technologies
Microarray Reagents
The PathDetect cis-reporting systems is designed for rapid and convenient assessment of the in vivo activation of these pathways. Each cis-reporter plasmid contains the luciferase reporter gene driven by a basic promoter element (TATA box) joined to tandem repeats of various binding elements.
  • Identify pathway-specific transcription activation
  • Easier and faster than blotting or gel-shift assays
  • Sensitive, quantitative assays
When PathDetect cis-reporting and expression plasmids encoding the gene of interest are co-transfected into mammalian cells, increased reporter activity indicates transcription activation and involvement of your gene product in the pathway leading to the enhancer element.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty