Would you like to log in again?
PCR Polishing Kit, Agilent Technologies
Supplier: Agilent Technologies
200409 200409
ST200409EA 291.84 USD InStock
ST200409 CAST200409
PCR Polishing Kit, Agilent Technologies
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
PCR polishing kit increases blunt-end cloning efficiencies associated with PCR-generated fragments.
  • Generates blunt-ended DNA fragments in 30 minutes
  • More efficient than Klenow polishing
  • Optimizes blunt-end cloning efficiencies
PCR Polishing Kit enables an increase in blunt-ended cloning efficiencies associated with polymerase chain reaction (PCR)-generated fragments. It polishes the ends of the 3’ overhang extensions of polymerase-generated DNA fragments directly from PCR amplification and can completely fill in 5’ overhangs to generate blunt ends.
Dramatic increases in the population of blunt-end DNA fragments following polishing with Agilent’s PCR Polishing Kit enables significant increase in overall experimental efficiency associated with procedures utilizing blunt-ended ligation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty