Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP6 RNA Polymerase, Agilent Technologies

Supplier: Agilent Technologies
600151 600151
ST600151EA 395.83 USD InStock
CAST600151 ST600151
SP6 RNA Polymerase, Agilent Technologies
Nucleic Acid Reagents RNA Synthesis and Transcription
A DNA-dependent RNA polymerase with high sequence specificity for SP6 promoter sequences.

  • Probes for nucleic acid hybridization
  • Templates for in vitro translation
  • Substrates for RNA processing studies
  • Exon and intron mapping of genomic DNA

SP6 RNA Polymerase is a DNA-dependent RNA polymerase with high sequence specificity for SP6 promoter sequences. SP6 RNA polymerase synthesizes RNA 5' to 3' and incorporates  35S, 32P and 33P ribonucleotides. This enzyme is isolated from an overproducing recombinant E. coli clone. SDS-polyacrylamide gel electrophoresis analysis of the SP6 RNA Polymerase assures 99% purity.

Unit definition: One unit will catalyze the incorporation of 1 N mole of nucleoside triphosphate into an acid insoluble form in 60 minutes at 37 °C.;Reaction Conditions: 40 mM Tris-HCl (pH 8.0), 8 mM MgCl₂, 50 mM NaCl, 2 mM spermidine, 30 mM DTT, 400 µM of each rNTP, 1 µg DNA template and 20 units enzyme in a 25 µl volume. Incubate 60 minutes at 37 °C.

Caution: For research use only. Not for use in diagnostic procedures.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty