Would you like to log in again?
Bubble Humidifier Kit, Vyaire Medical
Supplier: Vyaire Medical
002620
59100-108CS 262.82 USD InStock
59100-108
Bubble Humidifier Kit, Vyaire Medical
Ventilators
These humidifier systems feature a dual-purpose reservoir with a snap-open trigger, which makes a round opening for secure attachment of a humidifier adapter.
  • Prefilled with sterile water
  • Latex-free
The oxygen pathway is designed to help prevent particulate from reaching oxygen delivery tubing. An audible relief valve warns if the patient’s tubing becomes occluded. They are designed for convenient shelf storage.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty