Would you like to log in again?
Angle Two™ Stereotaxic Instrument, Leica
Supplier: Leica
Angle Two™
39464601 39464610
10030-374EA 33819.96 USD InStock
10030-374 10030-376
Angle Two™ Stereotaxic Instrument, Leica
Clinical Diagnostic Supplies Clinical Instruments
Angle Two™ small animal stereotaxic instrument enables varying the angle of approach between surgeries without loss of accuracy. Target setting and path of approach are integrated with an onscreen atlas, either mouse or rat or both.
  • Virtual skull flat feature corrects for head tilt
  • No vernier to read or arithmetic, so risk of human error is greatly reduced
  • Manual fine drive for precise transit and positioning
  • Electrical 120 V
This instrument helps the surgeon to spend less time on each animal, with more accurate results, and vary the angle of approach to avoid confounding action-at-target with path-of-approach.
The neuroscientist can improve the rigor and interpretability of stereotaxic research without adding heavy calculations or risk of error to the protocol, and therefore design experiments with fewer animals. Installed encoders connect to computer allow it to keep track of electrode tip location once shown the position of bregma.
Delivery information: Includes configured computer with monitor, keyboard, mouse, PCI card and Angle Two™ software installed for mouse or rat.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty