Would you like to log in again?
pET Expression System, Agilent Technologies
Supplier: Agilent Technologies
211523 211623 211521 211521 211523 211621 211621 211623
95040-836EA 1123.75 USD InStock
CA95040-834 95040-836 95040-834 95040-838 CA95040-836 99900-848 CA95040-838 CA99900-848
pET Expression System, Agilent Technologies
Protein Expression Protein Expression Systems
The pET E. coli expression system is a widely used in vivo bacterial expression system due to the strong selectivity of the bacteriophage T7 RNA polymerase for its cognate promoter sequences, the high level of activity of the polymerase, and the high efficiency of translation mediated by the T7 gene.
  • High-level expression upon induction
  • Convenient cloning sites for optimal expression of cloned protein-coding sequence
  • BL21(DE3) and BL21(DE3)pLysS competent cells are provided
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty