Would you like to log in again?
SureGuide Custom Amplified CRISPR Libraries, Agilent Technologies
Supplier: Agilent Technologies
G7555A-030 G7555A-010 G7555A-060
76193-714EA 12727.91 USD InStock
76193-714 76193-710 76193-712
SureGuide Custom Amplified CRISPR Libraries, Agilent Technologies
Vectors, Plasmids and Libraries
The SureGuide CRISPR/Cas9 libraries enable exome-wide knock-down screening that obtains superior fidelity, reduced screening time, and improved functional results.
SureGuide custom amplified libraries are delivered as pre-amplified oligo pools that are ready-to-clone into lentiviral vector. The CRISPR custom amplified libraries are also pre-amplified with fixed end sequences, making them compatible with the pSGLenti vector backbone.
In the libraries the entire guide region can be customized, allowing the flexibility to target new regions, design your own genome-wide or targeted screens, and even explore alternative CRISPR applications. CRISPR Pooled gRNA libraries may be created and applications that use a human or mouse U6 promoter will benefit from a library-based cloning technique using our proprietary SureVector technology.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty