Would you like to log in again?
SureGuide Unamplified Custom CRISPR Libraries, Agilent Technologies
Supplier: Agilent Technologies
G7555B-025 G7555B-100 G7555B-050 G7555B-005
76193-718EA 6299.66 USD InStock
76193-718 76193-716 76193-720 76193-722
SureGuide Unamplified Custom CRISPR Libraries, Agilent Technologies
Vectors, Plasmids and Libraries
The SureGuide CRISPR/Cas9 libraries enable exome-wide knock-down screening that obtains superior fidelity, reduced screening time, and improved functional results.
For full flexibility, also offers fully custom pooled CRISPR libraries where the promoter, cloning strategy, and entire scaffold can be synthesized to your specifications. These custom libraries come in kits containing all of the reagents necessary to amplify the pool and generate a clonable CRISPR library.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty