Would you like to log in again?
VariFlex, Agilent Technologies
Supplier: Agilent Technologies
240105 240105
99900-968EA 367.88 USD InStock
99900-968 CA99900-968
VariFlex, Agilent Technologies
Protein Expression Protein Expression Systems
The VariFlex protein expression system offers vectors with unique tags that enhance the utility of E. coli as an expression host.
  • All three SET tags in every vector set
  • Gentle SBP purification tag generates high-yield, pure proteins
Delivery information: Includes tag offering are three different SET tags which increase protein solubility and the SBP purification tag for high-yield, pure proteins.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty