Would you like to log in again?
Cooley Suction Tube, OR-Grade, Sklar®
Supplier: Sklar
07-1850
76191-536EA 454.3 USD InStock
76191-536
Cooley Suction Tube, OR-Grade, Sklar®
Cannulae
The Sklar® Cooley Suction Tube is used during surgical procedures to remove excess bodily fluids in order to maintain a clear surgical field.
The tube is connected to a suction machine for a power source.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty