Would you like to log in again?
T4 DNA Polymerase, BioVision
Supplier: BioVision
M1211-100
76264-930EA 291.88 USD InStock
76264-930
T4 DNA Polymerase, BioVision
Catalyzes the 5'→3' synthesis of DNA from a single stranded, primed DNA template.
  • Generates blunt end DNA by filling in 5'-overhangs or/and removing 3'-overhangs
  • High fidelity due to strong 3'→5' exonuclease activity
  • Synthesis of labeled DNA probes by the replacement reaction
  • Site-specific mutagenesis via primer extension from oligonucleotides
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty