Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Easy-A Hi-Fi Cloning Enzymes, Agilent Technologies

Supplier: Agilent Technologies
Easy-A Hi-Fi
600402 600404 600642 600642 600400 600640 600400 600640 600404 600402
99902-388EA 536.72 USD InStock
CA97066-456 99902-388 CA99902-388 ST600400 99902-358 97066-456 CA99902-358 99902-390 CA99902-390 CAST600400
Easy-A Hi-Fi Cloning Enzymes, Agilent Technologies
Nucleic Acid Reagents Cloning Kits and Ligation
The only RT-PCR kit able to offer both TA/UA cloning and high fidelity in a single system.

  • Produces PCR products ready for high-throughput TA/UA cloning with greater than 95 % cloning efficiency
  • Preferentially adds a single 3'-A overhang;Accuracy level is six times greater than Taq
  • HotStart formulated with a combination of antibodies, preventing priming until stringent primer annealing temperatures are reached, reducing background and improving detection sensitivity
  • Amplifies targets up to 10 kb from plasmid DNA and up to 5 kb from genomic DNA
  • The only RT-PCR kit available with the capacity to provide both TA/UA cloning and high fidelity
  • The kit produces PCR products ready for high-throughput TA/UA cloning with greater than 95% cloning

The Agilent Easy-A high-fidelity PCR cloning enzyme is a proprietary thermostable DNA polymerase formulation specifically designed for improved cloning with the StrataClone PCR Cloning Kit or with the TOPO TA Cloning vector and other T-/U-vectors.

Easy-A combines the higher accuracy of a proofreading enzyme with the high cloning efficiency of Taq and increased specificity with added hot start antibody formulation. PCR products amplified with Easy-A can be cloned directly, without performing the additional steps typically required when amplifying with proofreading polymerases.

The Agilent Easy-A One-Tube RT-PCR Kit enables the fast and accurate use of Easy-A high-fidelity cloning enzymes in many research applications. Easy-A enzymes combine the superior accuracy of proofreading enzymes with the high cloning efficiency of Taq.

Using the Easy-A One-Tube RT-PCR Kit PCR products can be cloned directly, without performing additional steps typically required when amplifying with proofreading polymerases. This enhances efficiency and ensures robust and consistent performance.

Caution: For research use only. Not for use in diagnostic procedures.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty