Would you like to log in again?
PathDetect Trans-Reporting Systems, Agilent Technologies
Supplier: Agilent Technologies
219054 219005 219036 219031 219010 219002 219026 219026 219000 219062 219001 219050 219068 219040 219056 219086 219010 219042 219065 219005 219041 219065 219015 219053 219004
95040-800EA 744.72 USD InStock
95040-800 CA99900-612 CA99900-732 CA99900-630 99900-642 CA99900-632 99900-600 CA95040-798 CA99900-598 ST219005 CAST219005 CA99900-600 CA99900-644 CA95040-800 CA99900-602 CA95040-802 CA99900-624 CA99900-586 CA99900-642 CA99900-608 CA99900-604 CA99900-626 CA99900-606 CA99900-592 CA99900-590
PathDetect Trans-Reporting Systems, Agilent Technologies
Microarray Reagents
PathDetect trans-reporting systems allow you to determine if the protein or compound of interest directly or indirectly activates one of six transcription factors: c-Jun, Elk1, CREB, c-Fos, ATF2 or CHOP.
  • Identify pathway-specific transcription activation
  • Easier and faster than blotting or gel-shift assays
  • Sensitive, quantitative assays
Easily investigate whethe the protein or compound influences the activation of any of the six transcription factors offered and etermine the direct or indirect involvement of new gene products, growth factors, and drug candidates in specific pathways.
Delivery information: Contains of a reporter plasmid, a trans-activator plasmid and both positive and negative control plasmids.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty