Would you like to log in again?
DpnI, Restriction Enzymes, BioVision Inc.
Supplier: BioVision
M1236-2000
76265-836EA 210.53 USD InStock
76265-836
DpnI, Restriction Enzymes, BioVision Inc.
Nucleic Acid Reagents Restriction Enzymes
A DNA digesting restriction enzyme
  • Simple procedure
  • Fast and convenient
The DpnI restriction enzyme digests DNA. DNA purified from a dam+ E. coli strain will be a substrate for DpnI due to the adenine methylation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty