Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
ViraPack Transfection Kit, Agilent Technologies
Supplier: Agilent Technologies
ViraPack
200488 200488
99900-560EA 340.42 USD InStock
CA99900-560 99900-560
ViraPack Transfection Kit, Agilent Technologies
Transfection Reagents
The ViraPack Transfection Kit is based on a modified CaPO4 method of DNA transfection to introduce viral plasmids into HEK293 packaging cell lines with high efficiency.
  • Unprecedented efficiency with retroviral, adenoviral, and AAV systems
  • Outperforms liposome-mediated transfection reagents in HEK293 cells
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty