Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
DNA Ligation Kit, Agilent Technologies
Supplier: Agilent Technologies
203003 203003
99900-690EA 379.23 USD InStock
99900-690 CA99900-690
DNA Ligation Kit, Agilent Technologies
Nucleic Acid Reagents Cloning Kits and Ligation
The DNA Ligation Kit includes T4 DNA ligase for highly efficient ligations.
  • For both sticky-end and blunt-end ligations
  • Reagents guaranteed to be DNase free while providing optimal ligation efficiency
  • Includes a plasmid control and a lambda control
The kit is optimized for use with lambda arms and plasmid vectors.
Delivery information: Kit contains T4 DNA ligase (300 U) 10X ligase buffer plasmid control Lambda control 10 mM rATP.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty