Would you like to log in again?
RNAMaxx High Yield Transcription Kit, Agilent Technologies
Supplier: Agilent Technologies
200339 200339
ST200339EA 339.92 USD InStock
ST200339 CAST200339
RNAMaxx High Yield Transcription Kit, Agilent Technologies
Nucleic Acid Reagents RNA Synthesis and Transcription
Rapidly and easily produce high yields of RNA from a DNA template.
  • Produces transcripts from a wide range of DNA template sizes, up to 9 kb
  • RNA yield 10 to 30 times the yield produced by conventional methods
  • 1 μg DNA template is 100 μg plus RNA transcript
  • For use in RNAi, microinjection studies, RNA-binding protein studies, hybridization studies, and microarrays
The RNAMaxx high yield transcription kit produces large quantities of RNA quickly and easily from a DNA template. Just 1 µg of template can yield 80 to 100 µg or more of RNA transcript in a short incubation. This is 10 to 30 times the yield produced by any conventional in vitro transcription reaction. The kit uses T7 RNA polymerase to produce high yields of RNA in just two hours.
RNAMaxx is ideal for the construction of non-isotopically labeled RNA probes. A variety of modified nucleotides, including amino allyl- and biotin-labeled nucleotides, are easily incorporated. The resulting labeled RNA transcripts can be used as probes for microarrays, Northern blot analysis, or in situ hybridization.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty