Would you like to log in again?
Proto® SkyHook™ Hammer/Ratchet Kit, ORS Nasco
Supplier: ORS Nasco
577-PSHS
76191-238EA 155.75 USD InStock
76191-238
Proto® SkyHook™ Hammer/Ratchet Kit, ORS Nasco
Fall Protection Fall Protection Kits
The SkyHook™ switch connector tethers to the tool on a short lanyard and provides secure tool transfers, a free range of motion, and positive tool control at the work site.
  • Mechanism is made from Dupont Delrin®, stainless steel springs and brass rivets for strength, durability, and corrosion resistance
  • The attachment securely anchors the SkyHook™ to wrist, belt, apron, or station
  • Just "swipe to switch", hand-to-hand, hand-to-belt, person-to-person, or person-to-station; all transfers are the same
Delivery information: Kit includes hammer holder and SkyHook™ switch connector.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty