Would you like to log in again?
Adhesive Floor Mats, Purus International
Supplier: Purus
219415 PS3660-34B
76240-582CS 421.13 USD InStock
76240-582 76244-890
Adhesive Floor Mats, Purus International
Floor Mats Tacky Mats
The multi-layered mat removes dirt and dust from foot traffic and equipment wheels prior to their entry into a clean or controlled environment.
  • Each numbered layer is fully coated with a particle grabbing adhesive
Once the top layer is saturated, a quick and easy removal of that layer exposes the next layer and returns the mat to maximum efficiency.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty