Would you like to log in again?
Door Closer, Zoro
Supplier: Zoro
351-UH-TB-EN
76249-602EA 1266.19 USD InStock
76249-602
Door Closer, Zoro
Door Systems Door Closers
This specialized door closer is available for storm doors, fire doors, overhead doors, and glass doors.
  • Non-handed orientation
  • Hold open closing type
This unit is made of aluminum and ideal to be used with doors have 60 and 48" of interior and exterior width respectively.
Certifications: Meets UL-10C and UBC 7-2 (1997) Positive Pressure Fire Test, ANSI A156.4 Grade 1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty