Would you like to log in again?
HDMI USB Cameras, Scienscope International
Supplier: Scienscope International
CC-HDMI-CD2
76248-142EA 3023.15 USD InStock
76248-142
HDMI USB Cameras, Scienscope International
Cameras
HDMI/USB 2.0 camera is the next step in the evolution of powerful line of digital HD cameras for microscopy.
  • On Screen control bar for setting menu
  • Split screen live image/capted image comparison
  • Wireless mouse support and control
It can be used as a live image HD (1080p) camera to a monitor using an HDMI cable or as a USB 2.0 camera with USB cable connected to any computer or Windows Tablet.
Delivery information: Includes power supply, HDMI cable, USB cable, 8 GB SD card, wired mouse
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty