Would you like to log in again?
Plastic Bio-Clamps, Bioconnexx
Supplier: BioConnex
NG075WH NG300WH NG200WH NG150WH
76239-010EA 64.9 USD InStock
76239-010 76239-012 76239-006 76239-008
Plastic Bio-Clamps, Bioconnexx
Clamps General Purpose Clamps
The Bio Clamp's design and construction lend itself to increased heat dissipation during high temperature applications.
  • Designed to fit sanitary flanges
  • Tamper evident
  • ISO Class 7 Clean Room produced and packaged
Reducing heat in non-metallic ferrules reduces the chance of warping and allowing leaks to occur. It can be sterilized by either steam or gamma irradiation.
Certifications: Conforms to ISO 2853.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty