Would you like to log in again?
Ring Lights, Scienscope International
Supplier: Scienscope International
IL-LED-R2
76243-010EA 716.48 USD InStock
76243-010
Ring Lights, Scienscope International
Lights Ring Lights
Adjustable ring light with power supply.
  • Dual LED ring light design
  • UV Free white fuse light
Less reflection on specular surfaces. Fits all Scienscope microscope bodies (ring adapter may be applied) and MAC zoom lens.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty