Screws, McMaster Carr
Supplier: McMaster-Carr
91251A342 91251A551 91251A537 91251A580 91251A442 91251A626 91251A583 91251A624 91251A630 91251A550 91251A540 91251A385
76247-060PK 56.03 USD InStock
76247-060 76247-070 76247-064 76247-052 76247-062 76247-050 76247-072 76247-068 76247-056 76247-066 76247-054 76247-058
Screws, McMaster Carr
Fasteners Screws
These black-oxide steel screws are mildly corrosion resistant in dry environments.
Coarse threads are the industry standard. Fine and extra-fine threads are closely spaced to prevent loosening from vibration. They are not compatible with coarse threads.
Certifications: Screws meets ASTM A574, ASTM A574M and ISO 898-1 comply with specifications and testing requirements for material quality.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty