Would you like to log in again?
4–in–1 Environmental Meter
Supplier: NEW GENIE GROUP, INC MS
45170
470310-240EA 266.7 USD InStock
470310-240
4–in–1 Environmental Meter
Weather Stations
Pocket-size rugged meter for Humidity, Temperature, Air Velocity and Light.
  • Air velocity range 80 to 5910 ft./min.
  • Light range 0 to 20000 lux
  • ±3% velocity accuracy
  • Ergonomic pocket-sized housing
  • 10 to 95% RH with fast response
  • 32 to 122 °F using thermistor
This 4-in-1 meter measures Humidity, Temperature, Air Velocity and Light. The dual LCD simultaneously displays Temperature and Air Velocity or Relative Humidity. High Temperature air or surface measurements can be made using the built-in Type K input. Comes complete with built-in Humidity sensor, Light sensor and Vane wheel, wrist strap, and 9-Volt battery.
Delivery information: Supplied with built-in humidity sensor, vane wheel, wrist strap and 9V battery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty