Would you like to log in again?
Protective Film Tape, 3M, Tapecase
Supplier: 3M
31825C-10X300F
76248-366EA 197.25 USD InStock
76248-366
Protective Film Tape, 3M, Tapecase
Seals General Purpose Sealing Films
This is made of low density polyethylene that has a low-tack acrylic adhesive system to help provide temporary protection of mill finished stainless steel and aluminum coils.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty