Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LEMD1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CJ1
75929-570EA 518 USD InStock
75929-570
Anti-LEMD1 Rabbit Polyclonal Antibody
Antibodies
LEMD1 (LEM domain-containing1) is a member of cancer-testis gene family. LEMD1 is normally expressed only in testis. Six alternatively spliced forms of LEMD1 transcripts in normal testis have been identified, but only one of the six was expressed in colorectal cancers. LEMD1 may represent a promising target antigen for immunotherapy of colorectal cancers. In addition, LEMD1 and SPATA19 are putative cancer biomarkers and promising targets for active immunotherapy.

Anti-LEMD1 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 20 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LEMD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty