Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-Citalopram oxalate

Supplier: SELLECK CHEMICALS LLC MS
Synonyms: (S)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate

S4064-50MG
103543-382EA 126.15 USD InStock
103543-382
(S)-Citalopram oxalate
(S)-Citalopram oxalate
Escitalopram oxalate is a selective serotonin (5-HT) reuptake inhibitor (SSRI) with Ki of 0.89 nM.


Formula: C₂₀H₂₁FN₂O
MW: 414.43 g/mol
MDL Number: MFCD06407826
CAS Number: 219861-08-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty