Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Protein LoBind Tubes, Eppendorf
Supplier: Eppendorf
Protein LoBind
0030122240 0030122216
76289-498EA 235.05 USD InStock
76289-498 76289-496
Protein LoBind Tubes, Eppendorf
Tubes Immunology Tubes
Protein LoBind tubes are specifically designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
  • Optimized sample recovery for improved assay results
  • Free of surface coating (silicone) to minimize the risk of sample interference
  • Higher recovery rate of proteins
  • PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Precise lid sealing to minimize evaporation
These tubes are widely used in preparation or storage of protein, peptide or antibody samples, enzymatic assays, prevention of sample loss during storage of virus stock solutions, storage of cell suspensions.
Delivery information: Available in tube, microplate, and deepwell plate formats for easy-up scaling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty