Would you like to log in again?
High-Temperature Soft Silicone Rubber Sheets, McMaster-Carr
Supplier: McMaster-Carr
9010K11 9010K83
MSPP-9010K11EA 44.42 USD InStock
MSPP-9010K11 MSPP-9010K83
High-Temperature Soft Silicone Rubber Sheets, McMaster-Carr
Rubber Sheeting
High-temperature super-soft silicone rubber sheets.
  • For outdoors use only
Tensile strength: 10A sheets: 200 psi; 20A sheets: 300 psi; 30A sheets: 650 psi.
Temperature range: 10A-20A sheets: –40 to 300 °F; 30A sheets: –80 to 400 °F.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty