Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Ultra-Low Temperature Freezers, Revco™ RDE Series, Thermo Scientific
Supplier: Thermo Scientific
Revco®
RDE40086FV RDE60086EA RDE50086FA RDE40086LD RDE60086LD RDE60086MA RDE50086FD RDE60086LA RDE60086FD RDE60086MD RDE50086FV RDE60086ED RDE50086LV RDE60086MD RDE40086LV RDE30086FD RDE40086FA RDE30086FV RDE60086EA RDE60086LV RDE30086LD RDE60086FA RDE40086FD RDE30086LA RDE50086LA RDE60086MA RDE30086FA RDE60086ED RDE60086FV RDE40086LA RDE50086LD RDE30086LV
76318-390EA 26277.15 USD InStock
76318-390 76318-392 76318-394 76318-396 76318-430 76318-428 76318-406 76318-408 76407-208 76318-420 CA76407-212 76318-388 76318-400 76318-422 76407-206 76318-402 76318-424 CA76407-210 76318-404 76318-426 76318-418 CA76407-206 CA76407-208 76318-398 76318-432 76318-410 76318-434 76318-412 76407-210 76318-414 76407-212 76318-416
Ultra-Low Temperature Freezers, Revco™ RDE Series, Thermo Scientific
Freezers Ultra Low Temperature Freezers
Thermo Scientific™ Revco™ RDE series ultra-low freezers feature four upright models, maximizing storage capacity from 30000 up to 60000 2 ml vials. Sustainability features include natural refrigerants and water-blown foam, compliant with many sustainability standards.
  • Reduced energy usage
  • Simple user interface
  • Ergonomic design
  • Natural refrigerants
  • Four capacities available
  • Manual defrosts
  • Smart-Vue compatible
  • 5 year bumper to bumper warranty
Save up to 25% in energy usage and heat rejection compared with ultra-low freezers with conventional refrigerant. Designed for daily use, cabinet temperature recovers 20% faster after opening the door. A quiet sound output below 53 dBA, enables the freezer to come out of the hallway and into the laboratory. Meets validation protocols at –70 °C and –80 °C with strong peak variation and steady-state temperature variation. Natural hydrocarbon refrigerants reduce system operating stresses promoting outstanding reliability and worry-free sample protection.
Freezers feature a re-designed, capacitive touch-screen user interface with easy-to-use set-up wizard, event log, temperature mapping and data downloads via a built-in USB port. Four capacities (300, 400, 500 and 600 2" box) to accommodate a variety of storage needs and laboratory footprint specifications.
All models contain five inner doors allowing easy sample access without exposing the contents of the entire freezer to ambient conditions. Improved customer convenience with a new ergonomically designed handle, high contrast eye-level display, and comfortable easy-access shelf locations. On board temperature and event archive allows quick access to freezer information and nbsp; Export a secure USB report and eliminate the need for a chart recorder. Compatible with most racking systems including a full range of Thermo Scientific™ racks for boxes, microplates, Thermo Scientific™ Matrix™ and Nunc CryoBank™ tubes.
Security features include alarm settings and padlock-compatible doors. Manufactured in an award-winning, zero-waste to landfill facility. Lower HVAC costs as a result of less heat emission into the environment.
Certifications: Compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), the European Union's F-Gas and other sustainability standards. All 115 and 208-230 V models are cULus. 76318-412, 76318-418, 76318-424, 76318-430, 76318-414, 76318-420, 76318-426, 76318-432, 76407-210 and 76407-212 also comply to FDA Class II. The 230 V models are all CE marked, and 76318-416, 76318-422, 76318-428 and 76318-434 also comply to FDA Class II.
Ordering information: Accessories are sold separately; some accessories are included in package options.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty