Would you like to log in again?
VWR® Puritan pH Buffer Standards
Supplier: Avantor
Puritan®
974692GMP 974892GMP 974492GMP 974792GMP 974992GMP
71002-418CS 1355.17 USD InStock
71002-418 71009-306 71002-416 71002-414 71002-420
VWR® Puritan pH Buffer Standards
Buffers pH Calibration Buffers
These cGMP manufactured pH buffers and conductivity standards enable reliable and accurate monitoring, prompt process management, and regulatory compliance of critical biopharmaceutical manufacturing processes.
Certifications: Traceable to the National Institute of Standards and Technology (NIST).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty