Would you like to log in again?
Insulated Chore Coats, VF Workwear
Supplier: VF Imagewear
YC15GY-SM-0R YC15GY-XL-0R YC15GY-2L-0R YC15GY-MD-0R YC15GY-LG-0R
76124-376EA 110.46 USD InStock
76124-376 76124-368 76124-370 76124-372 76124-374
Insulated Chore Coats, VF Workwear
Workwear
FR modacrylic insulation quilted to FR twill lining with adjustable hook-and-loop closure on barrel sleeve hem and a concealed storm cuff.
  • Bi-swing, two-piece action back
  • Side vents
  • Arc rating: 37.5
  • CAT 3
  • 10 oz. walls blend
Self fabric collar with concealed snaps for an optional hood and zip front closure with a protective storm flap covering. Pockets include two lower patch pockets, a chest pocket with snap closure (left side) and an interior cellphone/utility pocket (left side of chest).
Certifications: Meet NFPA 2112, NFPA 70E, ASTM F1506 standards
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty