Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Short-Sleeve Polo Shirts, VF Workwear

Supplier: VF Imagewear
FT10NV-XL-00 FT10NV-SM-00 FT10NV-2L-00 FT10NV-LG-00 FT10NV-MD-00
76124-386EA 200.26 USD InStock
76124-386 76124-378 76124-380 76124-382 76124-384
Short-Sleeve Polo Shirts, VF Workwear
Workwear
High-low hem for additional comfort (back hem is 1 inch longer)

  • Fully finished ribbed collar
  • Tapered hemmed sleeves
  • Arc rating: 8.2 Ebt
  • CAT 2
  • 6.7 oz. Tecasafe Plus Knit

Pockets include mic clip pockets on both shoulders and a dual pen pocket (left sleeve). Also has three-button front placket with microphone loop at the bottom of the placket.

Certifications: Meet NFPA 2112, NFPA 70E, ASTM F1506 standards

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty