Would you like to log in again?
T-Cube® Oral Fluid Drug Test, Alere™
Supplier: Alere Toxicology
T-Cube®
T-CUBE10BUP T-CUBE6MB T-CUBE10MO T-CUBE8BUP T-CUBE9BUP T-CUBE6O
76321-932BX 366.91 USD InStock
76321-932 76321-930 76321-926 76321-936 76321-934 76321-928
T-Cube® Oral Fluid Drug Test, Alere™
Drug Tests
With numerous oral fluid drug test devices on the market, it is important to select the one best suited to meet particular setting and needs. The T-Cube® oral fluid drug test is intended for employment and insurance testing, and offers fast and convenient oral fluid detection for multiple drugs assays.
  • 6 to 10 drug configurations available for a fully customized oral fluid testing program
  • Saliva saturation indicator ensures sufficient specimen volume is collected
  • Eliminates the need for gender-specific observation and limits adulteration and tampering
  • Drug test results in 5 minutes
Oral fluid drug screening simplifies collection processes and protects against specimen tampering, allowing users to make informed decisions about substance abuse.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty