Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc oxide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Zinci oxidumZinc (II) oxide

14439-6X1KG
BJ14439-1KGCS 1148.93 USD InStock
BJ14439-1KG
Zinc oxide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Zinc oxide


Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1.975 °C
Density: 5.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
Order Now
Specification Test Results
Assay (calculated to ignited substance) 99.0 - 100.5 %
Loss on ignition (500°C) Max. 1.0 %
Alkalinely reacting substances complying
Arsenic (As) Max. 3 ppm
Cadmium (Cd) Max. 10 ppm
Iron (Fe) Max. 10 ppm
Lead (Pb) Max. 50 ppm
Lead complying
Iron and other heavy metals complying
Carbonate and color of the solution complying
Carbonates and substances insoluble in acids complying
Residual solvents complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty