Would you like to log in again?
Closure Systems and Patented Caps, TriForest
Supplier: TriForest
TECT-24 CPG-24 CPT-24 CPG-53 CPG-38 TECT-53
76320-438CS 173.46 USD InStock
76320-438 76320-444 76320-446 76320-434 76320-442 76320-430
Closure Systems and Patented Caps, TriForest
Closures Bottle Closures
TriForest closures and caps are autoclavable, reusable, and recyclable.
  • Gamma radiation compatible to provide 10⁻⁶ sterility assurance level (SAL)
  • Cap material is Certified Animal Derived Component-Free
  • Compliance with USP Class VI guidelines
TriForest bottle caps are made with FDA approved, high density polyethylene resin. They are designed to provide a liner-less, leak-proof seal that is the hallmark of TriForest closure systems. Each bottle neck is uniquely designed to provide assurance of a leak-proof container.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty