Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® X Microbalances

Supplier: VWR
VWR®
VWR-21XA VWR-6XC VWR-6.21XC VWR-6.21XA VWR-52XA VWR-52XC VWR-21.52XA VWR-53XC VWR-21XC VWR-53XA VWR-6XA VWR-21.52XC
76321-896EA 25061.36 USD InStock
76321-896 76321-882 76321-878 76321-900 76321-876 76321-898 76321-904 76458-484 76321-902 76458-482 76321-880 76321-906
VWR® X Microbalances
Balances and Scales
The VWR® X-Series Microbalances advanced processing system and continuous environmental monitoring allows for the highest level of measurement accuracy and repeatability.

  • Capacity from 6.1 to 53 g
  • Automatic level control
  • Motorized glass draft shield (XA models)
  • Hands-free operation via IR proximity sensors
  • Automatic, internal calibration mechanism
  • 5.7" color, resistive touch screen
  • Built-in antistatic ionizer (XA models)
  • Three door draft shield chamber for easy sample introduction
  • Three year warranty

Suitable for continuous monitoring of environmental conditions (temperature, humidity, pressure, and vibrations).

It's cutting edge functionalities and features take data management, measurement quality and ease-of-use to a new level.

Certifications: CE marked.

Ordering information: Onsite installation and training of equipment included with purchase (continental United States and Canada). Please refer to related information tab for more information. To allow for the VWR® U-Series to perform to the highest standard, please refer to 'Laboratory Conditions' document within related information tab. Drop-shipment only.

Packaging: Packaged on small pallet.

Caution: Handle with extreme care.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty