Would you like to log in again?
Ward's® Albino Axolotl Habitat Sets
Supplier: VWR
Ward's®
WARD470320-434 WARD470320-438
470320-438EA 114.49 USD InStock
470320-438 470320-434
Ward's® Albino Axolotl Habitat Sets
Specimens Live Specimens Animals - Living Material
Large embryos with a clear coating makes them ideal for observing development.
  • They exhibit 'neoteny', keeping their larval characteristics into adulthood
  • This amphibian never leaves the water
  • Able to regenerate most body parts
Ambystoma mexicanum are unique salamanders keep their gills and stay fully aquatic even into adulthood, making them ideal subjects for embryological and developmental studies.
Widely known for their regenerative abilities, which diminish (but don't disappear) as they age, axolotls are studied in the biomedical field to understand the genetic triggers for cellular repair. These specimens are laboratory reared, as they are nearly extinct in their native Mexico due to degradation of their natural environment.
Ordering information: May be restricted in some states, check with your state's Department of Agriculture for restrictions. If Egeria najas is restricted in your state, the set that includes Elodea densa may be an alternative.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Packaging: Set contains one albino axolotl, a package of blackworms for feeding, either elodea densa or egeria najas, and an aquatic habitat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty