Would you like to log in again?
Jacketed Filter Reactor Systems, 20 L, Air Motor, Chemglass
Supplier: Chemglass
CG-1958-F621
89209-344EA 59907.54 USD InStock
89209-344
Jacketed Filter Reactor Systems, 20 L, Air Motor, Chemglass
Laboratory Reactor Systems
Jacketed filter glass reactors feature autocenter stand.
  • Complete reactor system on open air, autocenter stand
  • 150 °C maximum temperature limit for filter base
  • Filter base eliminates material transfers and significantly reduces losses
Jacketed filter glass reactors can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty