Would you like to log in again?
Wellwash™ Microplate Washers, Thermo Scientific
Supplier: Thermo Scientific
5165050 5165040
76333-272EA 8888.35 USD InStock
76333-272 76333-274
Wellwash™ Microplate Washers, Thermo Scientific
The Wellwash™ is a reliable microplate washer for 96-well plates particularly useful for routine ELISA applications.The instrument has many safety features including an aerosol cover that prevents aerosolisation of infectous diseases; liquid level sensors are present in both wash and waste bottlesand a plate sensor recognises if plate is present.
  • Large, colour graphical display and keyboard for easy, convenient use
  • User interface is logic and intuitive for quick protocols development
  • Unpressurised bottles are safe and secure to use
  • Automatic rinse and prime features assure safe performance
It has washing volume of 50 to 1000 μl and priming and rinsing volume of 5 to 100 ml.
Delivery information: Supplied with 1×2 litre wash bottle, 1×2 litre waste bottle, 1×8 or 1×12 wash head, aerosol cover.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty